missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AARSD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554624
This item is not returnable.
View return policy
Description
Immunogen sequence: GGVVILHRKE GDSEFMNIIA NEIGSEETLL FLTVGDEKGG GLFLLAGPPA SVETLGPRVA EVLEGKGAGK KGRFQGKATK MSRRMEAQAL LQDYISTQSA KE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
AARSD1 functions in trans to edit the amino acid moiety from incorrectly charged tRNA(Ala).
Specifications
| AARSD1 | |
| Polyclonal | |
| Unconjugated | |
| AARSD1 | |
| 1110069E20Rik; 2310044P18Rik; AA589600; AARSD1; Alanyl-tRNA deacylase alaX; alanyl-tRNA editing protein Aarsd1; alanyl-tRNA synthetase domain containing 1; alanyl-tRNA synthetase domain-containing protein 1; Alax; AlaXp-II; MGC2744 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80755 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BTE6 | |
| AARSD1 | |
| Recombinant protein corresponding to Human AARSD1. Recombinant protein control fragment (Product #RP-93206). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction