missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AARSD1 Antibody (012), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90276F
This item is not returnable.
View return policy
Description
AARSD1 Monoclonal specifically detects AARSD1 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| AARSD1 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 80755 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunofluorescence | |
| 12 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| alanyl-tRNA editing protein Aarsd1, alanyl-tRNA synthetase domain containing 1, Alanyl-tRNA synthetase domain-containing protein 1, MGC2744 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human AARSD1 (Q9BTE6-2; Thr176-Glu525). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction