Learn More
Invitrogen™ AARS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121142
Description
Positive test controls include: HepG2, SKOV3, U-251MG, MCF7. The target is usually found in the following locations: Mitochondrion. Immunogen sequence: RRELLATVKM LQRRANTAIR KLQMGQAAKK TQELLERHSK GPLIVDTVSA ESLSVLVKVV RQLCEQAPST SVLLLSPQPM GKVLCACQVA QGAMPTFTAE AWALAVCSHM GGKAWGSRVV AQGTGSTTDL EAALSIAQTY ALSQL.
The protein encoded by this gene belongs to the class-II aminoacyl-tRNA synthetase family. Aminoacyl-tRNA synthetases play critical roles in mRNA translation by charging tRNAs with their cognate amino acids. The encoded protein is a mitochondrial enzyme that specifically aminoacylates alanyl-tRNA. Mutations in this gene are a cause of combined oxidative phosphorylation deficiency 8. Catalyzes the attachment of alanine to tRNA(Ala) in a two-step reaction: alanine is first activated by ATP to form Ala-AMP and then transferred to the acceptor end of tRNA(Ala). Also edits incorrectly charged tRNA(Ala) via its editing domain.
Specifications
| AARS2 | |
| Polyclonal | |
| Unconjugated | |
| AARS2 | |
| AARS2; AARSL; alanine tRNA ligase 2, mitochondrial (putative); alanine--tRNA ligase, mitochondrial; alanine--tRNA ligase, mitochondrial {ECO:0000255; Alanyl-tRNA synthetase; alanyl-tRNA synthetase {ECO:0000255; alanyl-tRNA synthetase 2, mitochondrial; alanyl-tRNA synthetase 2, mitochondrial (putative); alanyl-tRNA synthetase like; alanyl-tRNA synthetase, mitochondrial; alaRS; alaRS {ECO:0000255; bA444E17.1; COXPD8; Gm89; HAMAP-Rule:MF_03133}; Kiaa1270; LKENP; MTALARS; MT-ALARS; probable alanyl-tRNA synthetase, mitochondrial | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57505 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5JTZ9 | |
| AARS2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 841-985 of human AARS2 (NP_065796.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.