missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AARS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566387
This item is not returnable.
View return policy
Description
Immunogen sequence: QLSNGTTRLLA VTGEQAQQAR ELGQSLAQEV KAATERLSLG SRDVAEALRL SKDIGRLIEA VETAVMP Highest antigen sequence identity to the following orthologs - mouse 75%, rat 75%.
The protein encoded by this gene belongs to the class-II aminoacyl-tRNA synthetase family. Aminoacyl-tRNA synthetases play critical roles in mRNA translation by charging tRNAs with their cognate amino acids. The encoded protein is a mitochondrial enzyme that specifically aminoacylates alanyl-tRNA. Mutations in this gene are a cause of combined oxidative phosphorylation deficiency 8. Catalyzes the attachment of alanine to tRNA(Ala) in a two-step reaction: alanine is first activated by ATP to form Ala-AMP and then transferred to the acceptor end of tRNA(Ala). Also edits incorrectly charged tRNA(Ala) via its editing domain.
Specifications
| AARS2 | |
| Polyclonal | |
| Unconjugated | |
| AARS2 | |
| AARS2; AARSL; alanine tRNA ligase 2, mitochondrial (putative); alanine--tRNA ligase, mitochondrial; alanine--tRNA ligase, mitochondrial {ECO:0000255; Alanyl-tRNA synthetase; alanyl-tRNA synthetase {ECO:0000255; alanyl-tRNA synthetase 2, mitochondrial; alanyl-tRNA synthetase 2, mitochondrial (putative); alanyl-tRNA synthetase like; alanyl-tRNA synthetase, mitochondrial; alaRS; alaRS {ECO:0000255; bA444E17.1; COXPD8; Gm89; HAMAP-Rule:MF_03133}; Kiaa1270; LKENP; MTALARS; MT-ALARS; probable alanyl-tRNA synthetase, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57505 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5JTZ9 | |
| AARS2 | |
| Recombinant Human AARS2. Recombinant protein control fragment (Product #RP-107060). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction