Learn More
Invitrogen™ AAR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116837
Description
Positive Samples: MCF7, A-549 Immunogen sequence: MAAVQMDPEL AKRLFFEGAT VVILNMPKGT EFGIDYNSWE VGPKFRGVKM IPPGIHFLHY SSVDKANPKE VGPRMGFFLS LHQRGLTVLR WSTLREEVDL SPAPESEVEA MRANLQELDQ FLGPYPYATL KKWISLTNFI SEATVEKLQP ENRQICAFSD VLPVLSMKHT KDRVGQNLPR CGIECKSYQE GLARLPEMKP RAGTEIRFSE LPTQMFPEGA TPAEITKHSM DLSYALETVL NKQFPSSPQD.
AAR2 encodes the homolog of the yeast A1-alpha2 repressin protein that is involved in mRNA splicing. Alternately spliced transcript variants have been found for this gene.
Specifications
| AAR2 | |
| Polyclonal | |
| Unconjugated | |
| AAR2 | |
| 0610011L14Rik; Aar2; AAR2 splicing factor homolog; AAR2 splicing factor homolog (S. cerevisiae); C20orf4; CGI-23; novel transcript; PRO0225; Protein AAR2 homolog; RGD1311066 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25980, 68295 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.23 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9D2V5, Q9Y312 | |
| AAR2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human AAR2 (NP_0563262). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.