missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35843-20ul
This item is not returnable.
View return policy
Description
AAR2 Polyclonal antibody specifically detects AAR2 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| AAR2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| AAR2 splicing factor homolog (S. cerevisiae), bA234K24.2, C20orf4, CGI-23, chromosome 20 open reading frame 4, DKFZp564N1363, hypothetical protein LOC25980 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human AAR2 (NP_056326.2).,, Sequence:, MAAVQMDPELAKRLFFEGATVVILNMPKGTEFGIDYNSWEVGPKFRGVKMIPPGIHFLHYSSVDKANPKEVGPRMGFFLSLHQRGLTVLRWSTLREEVDLSPAPESEVEAMRANLQELDQFLGPYPYATLKKWISLTNFISEATVEKLQPENRQICAFSDVLPVLSMKHTKDRVGQNLPRCGIECKSYQEGLARLPEMKPRAGTEIRFSELPTQMFPEGATPAEITKHSMDLSYALETVLNKQFPSSPQD | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 25980 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction