missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AAQ Chloride
GREENER_CHOICE

Product Code. p-7112887
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18736638

Brand: Tocris Bioscience™ 5462/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Photoswitchable Kv channel blocker

AAQ chloride is a photoswitchable Kv channel blocker (IC50 values are 2 and 64 μM at 500 nm and 380 nm respectively). Switches conformation from cis to trans at 500 nm and trans to cis at 380 nm. Exhibits minimal activity at Nav1.2 and L-type Ca2+ channels. Stimulates action potential firing of hippocampal neurons in vitro at 500 nm and restores visual responsiveness in blind mice at 380 nm.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N,N-Triethyl-2-oxo-2-[[4-[2-[4-[(1-oxo-2-propen-1-yl)amino]phenyl]diazenyl]phenylamino]ethanaminium chloride
Assay Percent Range >98%
Quantity 10 mg
Solubility Soluble to 20mM in DMSO
Molecular Formula C23H30ClN5O2
Formula Weight Observed MW: 443.97

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.