missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AAQ Chloride

Description
AAQ chloride is a photoswitchable Kv channel blocker (IC50 values are 2 and 64 μM at 500 nm and 380 nm respectively). Switches conformation from cis to trans at 500 nm and trans to cis at 380 nm. Exhibits minimal activity at Nav1.2 and L-type Ca2+ channels. Stimulates action potential firing of hippocampal neurons in vitro at 500 nm and restores visual responsiveness in blind mice at 380 nm.
Specifications
Specifications
| Chemical Name or Material | N,N,N-Triethyl-2-oxo-2-[[4-[2-[4-[(1-oxo-2-propen-1-yl)amino]phenyl]diazenyl]phenylamino]ethanaminium chloride |
| Assay Percent Range | >98% |
| Quantity | 10 mg |
| Solubility | Soluble to 20mM in DMSO |
| Molecular Formula | C23H30ClN5O2 |
| Formula Weight | Observed MW: 443.97 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?