missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AANAT Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565859
This item is not returnable.
View return policy
Description
Immunogen sequence: FLTLCPELSLG WFEEGCLVAF IIGSLWDKER LMQESLTLHR SGGHIAHLHV LAVHRAFRQQ GRGPILLWR Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.
Arylalkylamine N-Acetyltransferase (AANAT) is the penultimate enzyme is melatonin synthesis. Hormonal melatonin has been implicated in sleep and circadian clock function. Large changes in the abundance of AANAT are believed to be responsible for the large day/night rhythms in melatonin synthesis in the pineal gland and retina. Most research shows that AANAT activity is regulated by controlling the steady-state levels of the protein. Recent research, however, suggests that another mechanism may also play a role in controlling melatonin production without altering AANAT protein and activity. This mechanism may explain the sudden physiological increase in circulating melatonin observed at the onset of the dark period in humans.
Specifications
| AANAT | |
| Polyclonal | |
| Unconjugated | |
| Aanat | |
| Aanat; AA-NAT; Aralkylamine N-acetyltransferase; arylakylamine N-acetyltransferase; Arylalkylamine N - acetyltransferase (Serotonin N - acetyltransferase); arylalkylamine N-acetyltransferase; DSPS; Nat-2; Nat4; Seretonin N-acetyltransferase; serotonin acetylase; Serotonin N-acetyltransferase; SNAT | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16613 | |
| Aanat | |
| Recombinant Human Serotonin N-AT. Recombinant protein control fragment (Product #RP-106527). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction