missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AAMP Recombinant Rabbit Monoclonal Antibody (K01_1M25)
GREENER_CHOICE

Product Code. 30828462
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828462 100 μL
30825886 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828462 Supplier Invitrogen™ Supplier No. MA563278
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

AAMP Recombinant Monoclonal Antibody for Western Blot

The gene product is an immunoglobulin-type protein. It is found to be expressed strongly in endothelial cells, cytotrophoblasts, and poorly differentiated colon adenocarcinoma cells found in lymphatics. The protein contains a heparin-binding domain and mediates heparin-sensitive cell adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AAMP
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1M25
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene Aamp
Gene Accession No. Q13685
Gene Alias AAMP; Aamp-rs; Alternative macrophage activation-associated CC chemokine 1; AMAC1; AMAC-1; angio associated migratory cell protein; angio-associated migratory cell protein; angio-associated migratory protein; angio-associated, migratory cell protein; AU040907; CC chemokine ligand 18; CC chemokine PARC; C-C motif chemokine 18; C-C motif chemokine ligand 18; CCL18; CCL18(1-68); CCL18(3-69); CCL18(4-69); chemokine (C-C motif) ligand 18 (pulmonary and activation-regulated); chemokine (C-C), dendritic; CKb7; DCCK1; DC-CK1; Dendritic cell chemokine 1; macrophage inflammatory protein 4; MIP4; MIP-4; PARC; pulmonary and activation-regulated chemokine; SCYA18; small inducible cytokine A18; small inducible cytokine subfamily A (Cys-Cys), member 18, pulmonary and activation-regulated; small-inducible cytokine A18
Gene Symbols Aamp
Host Species Rabbit
Immunogen A synthetic peptide of human AAMP.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14, 227290, 301512
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.