missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AAMP Recombinant Rabbit Monoclonal Antibody (23GB1885), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554139
This item is not returnable.
View return policy
Description
The gene product is an immunoglobulin-type protein. It is found to be expressed strongly in endothelial cells, cytotrophoblasts, and poorly differentiated colon adenocarcinoma cells found in lymphatics. The protein contains a heparin-binding domain and mediates heparin-sensitive cell adhesion.
Specifications
| AAMP | |
| Recombinant Monoclonal | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13685 | |
| Aamp | |
| A synthesized peptide derived from human AAMP (15-51AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1885 | |
| Unconjugated | |
| Aamp | |
| AAMP; Aamp-rs; Alternative macrophage activation-associated CC chemokine 1; AMAC1; AMAC-1; angio associated migratory cell protein; angio-associated migratory cell protein; angio-associated migratory protein; angio-associated, migratory cell protein; AU040907; CC chemokine ligand 18; CC chemokine PARC; C-C motif chemokine 18; C-C motif chemokine ligand 18; CCL18; CCL18(1-68); CCL18(3-69); CCL18(4-69); chemokine (C-C motif) ligand 18 (pulmonary and activation-regulated); chemokine (C-C), dendritic; CKb7; DCCK1; DC-CK1; Dendritic cell chemokine 1; macrophage inflammatory protein 4; MIP4; MIP-4; PARC; pulmonary and activation-regulated chemokine; SCYA18; small inducible cytokine A18; small inducible cytokine subfamily A (Cys-Cys), member 18, pulmonary and activation-regulated; small-inducible cytokine A18 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14, 227290, 301512 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction