missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAMP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21220-1-AP-20UL
This item is not returnable.
View return policy
Description
The gene product is an immunoglobulin-type protein. It is found to be expressed strongly in endothelial cells, cytotrophoblasts, and poorly differentiated colon adenocarcinoma cells found in lymphatics. The protein contains a heparin-binding domain and mediates heparin-sensitive cell adhesion.Specifications
| AAMP | |
| Polyclonal | |
| Unconjugated | |
| AAMP | |
| AAMP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14, 227290, 301512 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13685 | |
| Aamp | |
| AAMP Fusion Protein Ag15655 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction