missing translation for 'onlineSavingsMsg'
Learn More

AAMP Antibody (2H2), Novus Biologicals™

Product Code. 18383208 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383208 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383208 Supplier Novus Biologicals Supplier No. H00000014M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AAMP Monoclonal antibody specifically detects AAMP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen AAMP
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 2H2
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001078
Gene Alias angio-associated migratory cell protein, angio-associated, migratory cell protein
Host Species Mouse
Immunogen AAMP (NP_001078.2, 111 a.a. ~ 210 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GEDDKAFVWRLSDGELLFECAGHKDSVTCAGFSHDSTLVATGDMSGLLKVWQVDTKEEVWSFEAGDLEWMEWHPRAPVLLAGTADGNTWMWKVPNGDCKT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.