missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AAMDC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558164
This item is not returnable.
View return policy
Description
Immunogen sequence: EIASLSWGQM KVKGSNTTYK DCKVWPGGSR TWDWRETGTE HSPGVQPADV KEVVEK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
AAMDC (C11orf67) may play a role in preadipocyte differentiation and adipogenesis.
Specifications
| AAMDC | |
| Polyclonal | |
| Unconjugated | |
| AAMDC | |
| 1810020D17Rik; 1810037D19Rik; Aamdc; adipogenesis associated Mth938 domain containing; adipogenesis associated Mth938 domain-containing protein; adipogenesis associated, Mth938 domain containing; C11orf67; C29H11orf67; CK067; hypothetical protein LOC553802; LI2; Mth938 domain-containing protein; PTD015; RGD1561459; Unknown (protein for MGC:133476); UPF0366 protein C11orf67; UPF0366 protein C11orf67 homolog; zgc:112239 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 28971 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H7C9 | |
| AAMDC | |
| Recombinant protein corresponding to Human AAMDC. Recombinant protein control fragment (Product #RP-97281). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction