Learn More
Masterflex Aalborg GFMS-010027 Compact Gas Mass Flowmeter, 0-5 LPM, Air/N2, Stainless Steel Body
Brand:  Masterflex 32648-66
Aalborg GFMS-010027 Compact Gas Mass Flowmeter, 0-5 LPM, Air/N2, Stainless Steel Body
Product Code. 17139018
  • ONLINE EXCLUSIVE PRICE
    £1295.00
    £999.00 / Each
    Save £296.00
    22% Off
Only null left
Estimated Shipment: 17-02-2026