missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAK1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-45214
This item is not returnable.
View return policy
Description
AAK1 Polyclonal specifically detects AAK1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| AAK1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Adaptor-associated kinase 1, AP2 associated kinase 1, AP2-associated protein kinase 1, DKFZp686F03202, DKFZp686K16132, EC 2.7.11.1, FLJ23712, FLJ25931, FLJ31060, FLJ45252, KIAA1048FLJ42882, MGC138170, MGC164568, MGC164570 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody recognizes and epitope within the internal region. Cross reacts with Human. Expected from sequence similarity: Pig and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_055726.3 | |
| AAK1 | |
| Peptide with sequence C-GSPRTSQQNVYNPSE corresponding to internal region according to NP_055726.3. | |
| 0.1 mg | |
| Protein Kinase | |
| 22848 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction