missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AAGAB Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA585464

Product Code. 16312405

  • £442.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Keep as concentrated solution. Predicted reactivity: Mouse (81%), Rat (83%), Bovine (87%). Positive Control: 293T, A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene interacts with the gamma-adaptin and alpha-adaptin subunits of complexes involved in clathrin-coated vesicle trafficking. Mutations in this gene are associated with type I punctate palmoplantar keratoderma. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AAGAB
Polyclonal
Unconjugated
AAGAB
2310007F21Rik; AAGAB; alpha- and gamma-adaptin binding protein; alpha- and gamma-adaptin-binding protein p34; KPPP1; p34; PPKP1; PPKP1A
Rabbit
Antigen affinity chromatography
RUO
79719
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry, Western Blot
0.79 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q6PD74
AAGAB
Recombinant protein encompassing a sequence within the center region of human FLJ11506. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.