missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AADAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583374
This item is not returnable.
View return policy
Description
Immunogen sequence: LSWLKQLQIK LHNPPTIHYP PSQGQMDLCV TSGSQQGLCK VFEMIINPGD NVLLDEPAYS GTLQSLHPLG CNIINVASDE SGIVPD Highest antigen sequence indentity to the following orthologs: Mouse - 71%, Rat - 71%.
This gene encodes a protein that is highly similar to mouse and rat kynurenine aminotransferase II. The rat protein is a homodimer with two transaminase activities. One activity is the transamination of alpha-aminoadipic acid, a final step in the saccaropine pathway which is the major pathway for L-lysine catabolism. The other activity involves the transamination of kynurenine to produce kynurenine acid, the precursor of kynurenic acid which has neuroprotective properties. Two alternative transcripts encoding the same isoform have been identified, however, additional alternative transcripts and isoforms may exist.
Specifications
| AADAT | |
| Polyclonal | |
| Unconjugated | |
| Aadat | |
| 2-aminoadipate aminotransferase; 2-aminoadipate transaminase; AadAT; Aadt; AI875679; alpha-aminoadipate aminotransferase; aminoadipate aminotransferase; KAT/AadAT; Kat2; KATII; KYAT2; kynurenine aminotransferase 2; kynurenine aminotransferase II; kynurenine/alpha-aminoadipate aminotransferase, mitochondrial; kynurenine--oxoglutarate; Kynurenine--oxoglutarate aminotransferase II; kynurenine--oxoglutarate transaminase 2; Kynurenine--oxoglutarate transaminase II; L kynurenine/alpha aminoadipate aminotransferase; mKat-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51166 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N5Z0 | |
| Aadat | |
| Recombinant protein corresponding to Human AADAT. Recombinant protein control fragment (Product #RP-96623). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction