Learn More
Invitrogen™ AADAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532646
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset (100%); Monkey (88%); Dog, Elephant (82%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
This gene encodes a protein that is highly similar to mouse and rat kynurenine aminotransferase II. The rat protein is a homodimer with two transaminase activities. One activity is the transamination of alpha-aminoadipic acid, a final step in the saccaropine pathway which is the major pathway for L-lysine catabolism. The other activity involves the transamination of kynurenine to produce kynurenine acid, the precursor of kynurenic acid which has neuroprotective properties. Two alternative transcripts encoding the same isoform have been identified, however, additional alternative transcripts and isoforms may exist.
Specifications
| AADAT | |
| Polyclonal | |
| Unconjugated | |
| Aadat | |
| 2-aminoadipate aminotransferase; 2-aminoadipate transaminase; AadAT; Aadt; AI875679; alpha-aminoadipate aminotransferase; aminoadipate aminotransferase; KAT/AadAT; Kat2; KATII; KYAT2; kynurenine aminotransferase 2; kynurenine aminotransferase II; kynurenine/alpha-aminoadipate aminotransferase, mitochondrial; kynurenine--oxoglutarate; Kynurenine--oxoglutarate aminotransferase II; kynurenine--oxoglutarate transaminase 2; Kynurenine--oxoglutarate transaminase II; L kynurenine/alpha aminoadipate aminotransferase; mKat-2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51166 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q8N5Z0 | |
| Aadat | |
| Synthetic 17 amino acid peptide from internal region of human AADAT. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.