missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AADAC Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99385CL1
This item is not returnable.
View return policy
Description
AADAC Polyclonal antibody specifically detects AADAC in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| AADAC | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| arylacetamide deacetylase, arylacetamide deacetylase (esterase), CES5A1, DACEC 3.1.1.3, EC 3.1.1, EC 3.1.1.79 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human AADAC. | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 13 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction