missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AACS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570507
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rat: 93%; Zebrafish: 86%.
Activates acetoacetate to acetoacetyl-CoA. May be involved in utilizing ketone body for the fatty acid-synthesis during adipose tissue development.
Specifications
| AACS | |
| Polyclonal | |
| Unconjugated | |
| AACS | |
| 2210408B16Rik; aacs; AcAc-CoA ligase; acetoacetate-CoA ligase; acetoacetyl-CoA ligase; acetoacetyl-CoA synthetase; ACSF1; acyl-CoA synthetase family member 1; homolog of C. elegans supressor of ras 5 (sur-5); protein sur-5 homolog; si:dkey-215k6.2; SUR5; SUR-5; zgc:56105 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 78894 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9D2R0 | |
| AACS | |
| synthetic peptide directed towards the middle region of human AACS (aa 481-530). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction