missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AACS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563617
This item is not returnable.
View return policy
Description
Immunogen sequence: RVVGYLPNSE HAVEAMLAAA SIGAIWSSTS PDFGVNGVLD RFSQIQPKLI FSVEAVVYNG KEHNHMEKLQ QVVKGLPDLK KVVVI Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
Activates acetoacetate to acetoacetyl-CoA. May be involved in utilizing ketone body for the fatty acid-synthesis during adipose tissue development.
Specifications
| AACS | |
| Polyclonal | |
| Unconjugated | |
| AACS | |
| 2210408B16Rik; aacs; AcAc-CoA ligase; acetoacetate-CoA ligase; acetoacetyl-CoA ligase; acetoacetyl-CoA synthetase; ACSF1; acyl-CoA synthetase family member 1; homolog of C. elegans supressor of ras 5 (sur-5); protein sur-5 homolog; si:dkey-215k6.2; SUR5; SUR-5; zgc:56105 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 65985 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86V21 | |
| AACS | |
| Recombinant protein corresponding to Human AACS. Recombinant protein control fragment (Product #RP-103342). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction