missing translation for 'onlineSavingsMsg'
Learn More

AAAS Antibody (OTI5B10), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18402087 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402087 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402087 Supplier Novus Biologicals Supplier No. NBP271358AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AAAS Monoclonal antibody specifically detects AAAS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen AAAS
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5B10
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias AAA, AAASb, achalasia, adrenocortical insufficiency, alacrimia, achalasia, adrenocortical insufficiency, alacrimia (Allgrove, triple-A), ADRACALA, Adracalin, ALADIN, Allgrove, triple-A, DKFZp586G1624
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 322-546 of human AAAS (NP_056480) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8086
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.