missing translation for 'onlineSavingsMsg'
Learn More

A939572, MedChemExpress

Product Code. p-200098784
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488088 10 mg 10mg
16410558 5 mg 5mg
16410356 2 mg 2mg
16484007 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16488088 Supplier Medchem Express Supplier No. HY50709

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A939572 is a potent, and orally bioavailable stearoyl-CoA desaturase1 (SCD1) inhibitor with IC50 values of mSCD1 and hSCD1, respectively.

A939572 is a potent, and orally bioavailable stearoyl-CoA desaturase1 (SCD1) inhibitor with IC50 values of mSCD1 and hSCD1, respectively.

Specifications

Chemical Name or Material A939572
CAS 1032229-33-6
Color White
Purity Grade Notes Research
Molecular Formula C20H22ClN3O3
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (257.82 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(N1CCC(OC2=C(Cl)C=CC=C2)CC1)NC3=CC=CC(C(NC)=O)=C3
Molecular Weight (g/mol) 387.86
Formula Weight 387.86
Percent Purity 99.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.