missing translation for 'onlineSavingsMsg'
Learn More

A77 1726 (E/Z) Mixture, TRC

Product Code. 30336414
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336414 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336414 Supplier Toronto Research Chemicals Supplier No. TRCA771720100MG
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Cyano-3-hydroxy-N-(4'-trifluoromethylphenyl)-crotone amide
CAS 108605-62-5
Molecular Formula C12 H9 F3 N2 O2
Quantity 100 mg
Synonym 2-Butenamide, 2-cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]-, 2-Cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]-2-butenamide, 2-Cyano-3-hydroxy-N-(4-trifluoromethylphenyl)crotonamide, 2-Cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]but-2-enamide, A77 1726 (E/Z) Mixture, (EZ)-Teriflunomide, 2-Cyano-3-hydroxy-N-(4'-trifluoromethylphenyl)-crotone amide
InChI Formula InChI=1S/C12H9F3N2O2/c1-7(18)10(6-16)11(19)17-9-4-2-8(3-5-9)12(13,14)15/h2-5,18H,1H3,(H,17,19)
SMILES CC(=C(C#N)C(=O)Nc1ccc(cc1)C(F)(F)F)O
IUPAC Name 2-cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]but-2-enamide
Molecular Weight (g/mol) 270.21
Formula Weight 270.0616
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.