missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A4GNT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22670-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein from the glycosyltransferase 32 family. The enzyme catalyzes the transfer of N-acetylglucosamine to core 2 branched O-glycans. It forms a unique glycan, GlcNAcalpha1->4Galbeta->R and is largely associated with the Golgi apparatus membrane.Specifications
| A4GNT | |
| Polyclonal | |
| Unconjugated | |
| A4GNT | |
| A4GNT, alpha4GnT | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 333424, 51146, 685758 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.44 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14BT6, Q9UNA3 | |
| A4gnt | |
| A4GNT Fusion Protein Ag18432 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction