Learn More
A4GNT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22670-1-AP-150UL
 View more versions of this product
Product Code. 16862035
  • £349.00 / 150µL
Description
This gene encodes a protein from the glycosyltransferase 32 family. The enzyme catalyzes the transfer of N-acetylglucosamine to core 2 branched O-glycans. It forms a unique glycan, GlcNAcalpha1->4Galbeta->R and is largely associated with the Golgi apparatus membrane.
Specifications
Specifications
A4GNT
Polyclonal
Unconjugated
A4GNT
A4GNT, alpha4GnT
Rabbit
Antigen Affinity Chromatography
RUO
333424, 51146, 685758
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.44 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q14BT6, Q9UNA3
A4gnt
A4GNT Fusion Protein Ag18432
150 μL
Primary
Human, Mouse, Rat