missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ A4GALT Monoclonal Antibody (38.13)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA544001
This item is not returnable.
View return policy
Description
This antibody recognizes CD77 (globotriaosylceramide Gb3), a neutral glycosphingolipid expressed mainly on the surface of B cell populations, such as germinal center B cells and Burkitt's lymphoma cells.
The protein encoded by this gene catalyzes the transfer of galactose to lactosylceramide to form globotriaosylceramide, which has been identified as the P(k) antigen of the P blood group system. The encoded protein, which is a type II membrane protein found in the Golgi, is also required for the synthesis of the bacterial verotoxins receptor.
Specifications
| A4GALT | |
| Monoclonal | |
| 1 mg/mL | |
| TBS with 15mM sodium azide; pH 8 | |
| Q9NPC4 | |
| A4galt | |
| Daudi cell line (Burkitts lymphoma). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry | |
| 38.13 | |
| Unconjugated | |
| A4galt | |
| A14GALT; A4GALT; A4galt {ECO:0000312; A4GALT1; alpha 1,4-galactosyltransferase; alpha 14-galactosyltransferase; alpha-1,4-galactosyltransferase; Alpha-1,4-N-acetylglucosaminyltransferase; alpha4Gal-T1; cd77; CD77 synthase; Gb3; Gb3 synthase; Gb3/CD77 synthase; Gb3S; Globotriaosylceramide synthase; lac; lactosylceramide 4-alpha-galactosyltransferase; P blood group (P one antigen); P(k); P(k) antigen synthase; P1; P1/Pk synthase; P1PK; PK; RGD:621583}; truncated alpha 1,4-galactosyltransferase; UDP-galactose:beta-D-galactosyl-beta1-R 4-alpha-D-galactosyltransferase | |
| Rat | |
| sequential chromatography | |
| RUO | |
| 53947 | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction