Description
The A375 Cell Lysate is derived from human. It has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Cell Line | A375 cell line lysate (amelanotic melanoma cell line) |
| Quantity | 0.1 mg |
| Storage Buffer | 1X SDS sample buffer: 60mM Tris-Hcl (pH 6.8) 10mM EDTA 2 % SDS 0.03% Bromophenol Blue 10% glycerol 0.3% Beta-mercaptoethanol |
| Content And Storage | -80°C in the dark. Avoid freeze-thaw cycles. |
| Product Line | Novus |