missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ A33 Monoclonal Antibody (1047587)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561256
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The glycoprotein encoded by this gene is a cell surface antigen that is expressed in greater than 95% of human colon cancers. The open reading frame encodes a 319-amino acid polypeptide having a putative secretory signal sequence and 3 potential glycosylation sites. The predicted mature protein has a 213-amino acid extracellular region, a single transmembrane domain, and a 62-amino acid intracellular tail. The sequence of the extracellular region contains 2 domains characteristic of the CD2 subgroup of the immunoglobulin (Ig) superfamily.
Specifications
| A33 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q99795 | |
| Gpa33 | |
| Mouse myeloma cell line NS0-derived recombinant human A33, Ile22-Val235. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry | |
| 1047587 | |
| Unconjugated | |
| Gpa33 | |
| 2010310L10Rik; 2210401D16Rik; A33; A33 antigen; BB116197; cell surface A33 antigen; Glycoprotein A33; glycoprotein A33 (transmembrane); GPA33; mA33; MGC129986; MGC129987; transmembrane glycoprotein A33 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 10223 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction