missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A33/GPA33 Antibody (SR1883), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £341.00
Specifications
| Antigen | A33/GPA33 |
|---|---|
| Clone | SR1883 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617734
|
Novus Biologicals
NBP3-22071-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623325
|
Novus Biologicals
NBP3-22071-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A33/GPA33 Monoclonal antibody specifically detects A33/GPA33 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
| A33/GPA33 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| A33, cell surface A33 antigen, Glycoprotein A33, glycoprotein A33 (transmembrane), MGC129986, MGC129987, transmembrane glycoprotein A33 | |
| A synthesized peptide derived from human A33/GPA33 (Uniprot #: Q99795) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1883 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 10223 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title