missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A2LD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23280-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene aids in the proteolytic degradation of crosslinked fibrin by breaking down isodipeptide L-gamma-glutamyl-L-epsilon-lysine, a byproduct of fibrin degradation. The reaction catalyzed by the encoded gamma-glutamylaminecyclotransferase produces 5-oxo-L-proline and a free alkylamine. Two transcript variants encoding the same protein have been found for this gene.Specifications
| A2LD1 | |
| Polyclonal | |
| Unconjugated | |
| GGACT | |
| A2LD1, AIG2 like domain 1, GGACT | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 223267, 290500, 87769 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q4KM86, Q923B0, Q9BVM4 | |
| Ggact | |
| A2LD1 Fusion Protein Ag18549 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction