missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ A2BP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103622
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FOX1.
Ataxin-2 binding protein 1 has an RNP motif that is highly conserved among RNA-binding proteins. This protein binds to the C-terminus of ataxin-2 and may contribute to the restricted pathology of spinocerebellar ataxia type 2 (SCA2). Ataxin-2 is the gene product of the SCA2 gene which causes familial neurodegenerative diseases. Ataxin-2 binding protein 1 and ataxin-2 are both localized in the trans-Golgi network. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene. Additional transcript variants have been found but their full length nature has not been determined.
Specifications
| A2BP1 | |
| Polyclonal | |
| Unconjugated | |
| Rbfox1 | |
| 2BP1; A FOX-1; A2BP; A2BP1; ataxin 2 binding protein 1; ataxin 2-binding protein 1; ataxin-2-binding protein 1; FOX1; fox-1; fox-1 homolog A; fox-1-like RNA-binding protein 1; hexaribonucleotide binding protein 1; hexaribonucleotide binding protein 1 isoform alpha; hexaribonucleotide binding protein 1 isoform beta; hexaribonucleotide-binding protein 1; HRNBP1; MNCb-3035; RBFOX1; RNA binding fox-1 homolog 1; RNA binding protein fox-1 homolog 1; RNA binding protein, fox-1 homolog (C. elegans) 1; RNA binding protein, fox-1 homolog 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 268859, 302920, 54715 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9JJ43, Q9NWB1 | |
| Rbfox1 | |
| A synthesized peptide derived from human RBFOX1(Accession Q9NWB1), corresponding to amino acid residues A22-H72. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction