missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ A26C2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122305
This item is not returnable.
View return policy
Description
Belongs to the POTE family. 3 isoforms of the human protein are produced by alternative splicing. Bera T.K., Gene 337:45-53(2004).
Specifications
| A26C2 | |
| Polyclonal | |
| Unconjugated | |
| POTEG | |
| A26C2; ACTBL1; ANKRD26-like family C member 2; ANKRD26-like family C, member 2; cancer/testis antigen family 104, member 4; CT104.4; POTE ankyrin domain family member G; POTE ankyrin domain family, member G; POTE14; POTE-14; POTE14alpha; POTE22; POTEG; Prostate, ovary, testis-expressed protein on chromosome 14; protein expressed in prostate, ovary, testis, and placenta 14 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 404785 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q6S5H5 | |
| POTEG | |
| Recombinant POTE Ankyrin Domain Family, Member G (POTEG), Leu177-Asp330 (Accession Q6S5H5). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction