missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ A26C2/A26C3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599693
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total POTE-14/22.
Belongs to the POTE family. 3 isoforms of the human protein are produced by alternative splicing. Bera T.K., Gene 337:45-53(2004).
Specifications
| A26C2/A26C3 | |
| Polyclonal | |
| Unconjugated | |
| POTEG | |
| 4921537P18Rik; 4930456F22Rik; A26C2; A26C3; ACTBL1; actin, beta-like 1; ANKRD26-like family C member 2; ANKRD26-like family C member 3; ANKRD26-like family C, member 2; ANKRD26-like family C, member 3; cancer/testis antigen family 104, member 4; cancer/testis antigen family 104, member 7; CT104.4; CT104.7; gonad specific ankyrin repeat protein; Gsarp1; Gsarp2; POTE ankyrin domain family member A; POTE ankyrin domain family member G; POTE ankyrin domain family member H; POTE ankyrin domain family, member G; POTE ankyrin domain family, member H; POTE14; POTE-14; POTE14alpha; POTE14B; POTE22; POTE-22; Poteg; POTEH; Prostate, ovary, testis-expressed protein on chromosome 14; prostate, ovary, testis-expressed protein on chromosome 22; protein expressed in prostate, ovary, testis, and placenta 14; protein expressed in prostate, ovary, testis, and placenta 22; protein expressed in prostate, ovary, testis, and placenta POTE14 like | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23784, 404785, 70952 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6S545, Q6S5H5 | |
| POTEG, POTEH | |
| A synthesized peptide derived from human POTEH(Accession Q6S545), corresponding to amino acid residues G479-E529. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction