missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ A20A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575795
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Ankyrins are membrane adaptor molecules that play important roles in coupling integral membrane proteins to the spectrin-based cytoskeleton network. Mutations of ankyrin genes lead to severe genetic diseases such as fatal cardiac arrhythmias and hereditary spherocytosis. ANKRD20A (ankyrin repeat domain-containing protein 20A) is an 823 amino acid protein that contains five ANK repeats. The gene encoding ANKRD20A maps to chromosome 9, which consists of about 145 million bases and encodes nearly 900 genes. Considered to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype. Hereditary hemorrhagic telangiectasia, which is characterized by harmful vascular defects, and familial dysautonomia are associated with chromosome 9. Also, chromosome 9 is partnered with chromosome 22 in the translocation leading to the aberrant production of BCR-ABL fusion protein often found in leukemias.
Specifications
| A20A1 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD20A1 | |
| ANKRD20A; ANKRD20A1; ankyrin repeat domain 20 family member A1; ankyrin repeat domain 20 family, member A1; ankyrin repeat domain 20A; ankyrin repeat domain-containing protein 20A1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84210 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5TYW2 | |
| ANKRD20A1 | |
| Synthetic peptide corresponding to residues in Human A20A1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction