missing translation for 'onlineSavingsMsg'
Learn More

A20/TNFAIP3 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18426034 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426034 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426034 Supplier Novus Biologicals Supplier No. NBP177024JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A20/TNFAIP3 Polyclonal antibody specifically detects A20/TNFAIP3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen A20/TNFAIP3
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias A20TNFA1P2, EC 3.4.19.12, EC 6.3.2.-, MGC104522, MGC138687, OTU domain-containing protein 7C, OTUD7CMGC138688, Putative DNA-binding protein A20, TNF alpha-induced protein 3, tumor necrosis factor alpha-induced protein 3, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha-induced protein 3, Zinc finger protein A20
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the center of human A20/TNFAIP3 . The immunogen is located within amino acids 340 - 390 of A20/TNFAIP3 Antibody. Amino Acid Squence: VQHEYKKWQENSDQARR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Inflammation, Innate Immunity, Necroptosis, Neurodegeneration, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7128
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.