missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ A1BG Monoclonal Antibody (D4), Invitrogen™

Mouse Monoclonal Antibody
£161.00 - £380.00
Specifications
| Antigen | A1BG |
|---|---|
| Clone | D4 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17999742
|
Invitrogen™
MA550884 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17900423
![]() |
Invitrogen™
MA550883 |
200 μL |
£380.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alpha-1-B glycoprotein (A1BG) which is a plasma protein with no known function belongs to the immunoglobulin superfamily. Although the human A1BG has been known for four decades, and the information about the amino acid sequence, chromosomal assignment, and even genetic polymorphism in different populations have been known, no biological function has been suggested. A1BG (Mr approximately equal to 63,000) consists of one polypeptide chain of 474 amino acids with four glucosamine oligosaccharides. The polypeptide has five intrachain disulfide bonds and consists of five repeating structural domains, each containing about 95 amino acids and one disulfide bond. Recently human cysteine-rich secretory protein 3 (CRISP-3) has been found to be a binding partner of A1B, suggesting the A1BG-CRISP-3 complex displays a protecting function from a potentially harmful effect of free CRISP-3 in circulation.Specifications
| A1BG | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| A1B; A1bg; A1bg {ECO:0000312; ABG; Alpha-1-B glycoprotein; alpha-1-B-glycoprotein; alpha-1B-glycoprotein; alpha1B-glycoprotein; C44; DKFZp686F0970; epididymis secretory sperm binding protein Li 163pA; GAB; HEL-S-163pA; HYST2477; Liver regeneration-related protein 1; RGD:69417} | |
| A1BG | |
| IgG1 κ | |
| Protein A/G | |
| A1BG |
| D4 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P04217 | |
| 1 | |
| Recombinant Human a1BG protein, Ala22-Ala206 (Accession #P04217). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title