missing translation for 'onlineSavingsMsg'
Learn More

A1874, MedChemExpress

Product Code. 16474808
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16474808

Brand: Medchem Express HY11430510mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A1874 is a nutlin-based (MDM2 ligand) and BRD4-degrading PROTAC with a DC50 of 32 nM (induce BRD4 degradation in cells). Effective in inhibiting many cancer cell lines proliferation.

A1874 is a nutlin-based (MDM2 ligand) and BRD4-degrading PROTAC with a DC50 of 32 nM (induce BRD4 degradation in cells). Effective in inhibiting many cancer cell lines proliferation.

Specifications

Chemical Name or Material A1874
CAS 2064292-12-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C58H62Cl3F2N9O7S
Quantity 10 mg
Solubility Information DMSO : 150 mg/mL (127.81 mM; Need ultrasonic)
SMILES ClC(C=C1)=CC=C1C2=N[C@H](C3=NN=C(C)N3C4=C2C(C)=C(C)S4)CC(NCCOCCOCCOCCNC(C5=CC(OC)=C(NC([C@H]6[C@H](C7=C(F)C(Cl)=CC=C7)[C@](C8=CC=C(Cl)C=C8F)(C#N)[C@H](CC(C)(C)C)N6)=O)C=C5)=O)=O
Molecular Weight (g/mol) 1173.59
Formula Weight 1173.59
Percent Purity 99.28%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.