missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A01

Description
A01 is a high affinity Smurf1 E3 ubiquitin ligase inhibitor (Kd = 3.7 nM). Attenuates Smurf1-mediated Smad 1/5 degradation and enhances BMP signaling. Potentiates BMP-2-induced osteoblastic activity in C2C12 myoblasts and MC3T3-E1 osteoblast precursor cells.
Specifications
Specifications
| Purity | >98% |
| Quantity | 10 mg |
| Product Type | A01 |
| Molecular Weight (g/mol) | 512.93 |
| Inhibitors | A01 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?