Learn More
SPEX Certiprep™ a-Terpineol
Single-Component Organic Standards
Brand: SPEX Certiprep™ S-4145
Code : NEW
Additional Details : CAS Number : 67-56-1 Weight : 0.01000kg Transport : UN number : 1230 Chem class : 3 Pack group : II
Description
- Single-component standards are available as stock items. They are supplied in methanol or in another solvent when appropriate.
- Additional solvents include methylene chloride, acetone, benzene, hexane, acetonitrile, THF, and water.
- Concentrations other than these, as well as custom matrices, are readily available, but a minimum order of five ampules may be required in some cases.
Safety and Handling
- ALPHA-TERPINEOL
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methanol