missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ A-RAF Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Packungsgröße:
0.1mL
Spezifikation
Spezifikation
| For Use With (Application) | Antibody Competition |
| Formulation | PBS and 1 M Urea, pH 7.4. |
| Gene ID (Entrez) | 369 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 25 kDa |
| Quantity | 0.1 mL |
| Storage Requirements | Store at -20°C. Avoid freeze/thaw cycles. |
| Gene Alias | ARAF1A-RAF, EC 2.7.11, EC 2.7.11.1, Oncogene ARAF1, Pk., PKS2A-Raf proto-oncogene serine/threonine-protein kinase, Proto-oncogene A-Raf, Proto-oncogene A-Raf-1, Proto-oncogene Pk., RAFA1, Ras-binding protein DA-Raf, serine/threonine-protein kinase A-Raf, v-raf murine sarcoma 3611 viral oncogene homolog, v-raf murine sarcoma 3611 viral oncogene homolog 1 |
| Gene Symbol | ARAF |
| Inhibitor Properties | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-49479. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL |
| Research Category | Protein Kinase, Signal Transduction |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur