Learn More
Eurofins DiscoverX A-Raf Protein, Active
Description
This proto-oncogene belongs to the RAF subfamily of the Ser/Thr protein kinase family, and maybe involved in cell growth and development. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
GenBank U01337.1 The recombinant protein contains the amino acid substitution Y301D and Y302D with reference to GenBank U01337.1. These mutations provide constitutive activation of the enzyme.
Specifications
Specifications
| Formulation | mg/mL of enzyme in 40 mM Tris/HCl pH 8.0, 200 mM NaCl, 0.08 mM EGTA, 0.024% Brij-35, 4 mM glutathione, 20% (v/v) glycerol, 0.8 mM benzamidine, 0.16 mM PMSF, 0.08% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 65 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal GST |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥47% |
| Protein | A-Raf, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.