missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX A-Raf Protein, Active

Product Code. p-7228359
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17320854

Brand: Eurofins DiscoverX 14956

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal GST-tagged, recombinant, human A-Raf, for use in Kinase Assays

This proto-oncogene belongs to the RAF subfamily of the Ser/Thr protein kinase family, and maybe involved in cell growth and development. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

GenBank U01337.1 The recombinant protein contains the amino acid substitution Y301D and Y302D with reference to GenBank U01337.1. These mutations provide constitutive activation of the enzyme.

Specifications

Formulation mg/mL of enzyme in 40 mM Tris/HCl pH 8.0, 200 mM NaCl, 0.08 mM EGTA, 0.024% Brij-35, 4 mM glutathione, 20% (v/v) glycerol, 0.8 mM benzamidine, 0.16 mM PMSF, 0.08% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 65 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal GST
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥47%
Protein A-Raf, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.