missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A-Myb Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-90171
This item is not returnable.
View return policy
Description
A-Myb Polyclonal specifically detects A-Myb in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| A-Myb | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| MYBL1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:EANAVLSSLQTIPEFAETLELIESDPVAWSDVTSFDISDAAASPIKSTPVKLMRIQHNEGAMECQFNVSLVLEGKKNTCNGGNSEAVPLTSPNIAKFSTPPAILRKKRKMRVGHSPGSELRDGSLNDGGN | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 4603 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.1mg/mL | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| A-MYB, AMYBMGC120059, MGC120061, Myb-like protein 1, myb-related protein A, v-myb avian myeloblastosis viral oncogene homolog-like 1, v-myb myeloblastosis viral oncogene homolog (avian)-like 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction