missing translation for 'onlineSavingsMsg'
Learn More

A-Myb Antibody (2A2), Novus Biologicals™

Product Code. 18378477 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378477 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378477 Supplier Novus Biologicals Supplier No. H00004603M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

A-Myb Monoclonal antibody specifically detects A-Myb in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen A-Myb
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2A2
Conjugate Unconjugated
Dilution Western Blot 1:100 to 1:2000, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. XP_034274
Gene Alias A-MYB, AMYBMGC120059, MGC120061, Myb-like protein 1, myb-related protein A, v-myb avian myeloblastosis viral oncogene homolog-like 1, v-myb myeloblastosis viral oncogene homolog (avian)-like 1
Host Species Mouse
Immunogen MYBL1 (XP_034274, 625 a.a. ~ 734 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KSLVLDNWEKEESGTQLLTEDISDMQSENRFTTSLLMIPLLEIHDNRCNLIPEKQDINSTNKTYTLTKKKPNPNTSKVVKLEKNLQSNCEWETVVYGKTEDQLIMTEQAR
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4603
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.