missing translation for 'onlineSavingsMsg'
Learn More

a-Hydroxy-3,4-bis(phenylmethoxy)benzeneacetic Acid Phenylmethyl Ester, TRC

Product Code. 30399308
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399308 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399308 Supplier Toronto Research Chemicals Supplier No. TRCH8301601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-Hydroxy-3,4-bis(phenylmethoxy)benzeneacetic Acid Phenylmethyl Ester
CAS 113026-97-4
Molecular Formula C29H26O5
Quantity 1 g
InChI Formula InChI=1S/C29H26O5/c30-28(29(31)34-21-24-14-8-3-9-15-24)25-16-17-26(32-19-22-10-4-1-5-11-22)27(18-25)33-20-23-12-6-2-7-13-23/h1-18,28,30H,19-21H2
SMILES OC(C1=CC(OCC2=CC=CC=C2)=C(C=C1)OCC3=CC=CC=C3)C(OCC4=CC=CC=C4)=O
IUPAC Name benzyl 2-(3,4-bis(benzyloxy)phenyl)-2-hydroxyacetate
Molecular Weight (g/mol) 454.51
Formula Weight 454.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.