missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A. aeolicus BPL/BioID2 Antibody (SS QD1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59940-0.025mg
This item is not returnable.
View return policy
Description
A. aeolicus BPL/BioID2 Monoclonal antibody specifically detects A. aeolicus BPL/BioID2 in Bacteria samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/ Immunofluorescence
Specifications
| A. aeolicus BPL/BioID2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Simple Western, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2 κ |
| Western Blot, Simple Western, Immunofluorescence | |
| SS QD1 | |
| Unconjugated | |
| PBS | |
| GST fused to A. aeolicus BPL/BioID2 | |
| 0.025 mg | |
| Epitope Tags | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction