missing translation for 'onlineSavingsMsg'
Learn More

a,a,a’,a’-Tetramethyl-5-methyl-1,3-benzenediacetonitrile, TRC

Product Code. 30423500
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423500 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423500 Supplier Toronto Research Chemicals Supplier No. TRCT303615100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2'-(5-Methylbenzene-1,3-diyl)bis(2-methylpropanenitrile)
CAS 120511-72-0
Molecular Formula C15 H18 N2
Quantity 100 g
Synonym 1,3-Benzenediacetonitrile','α,α,α',α',5-pentamethyl-, 2,2'-(5-Methyl-1,3-phenylene)di(2-methylpropionitrile), 2-[3-(Cyanodimethylmethyl)-5-methylphenyl]-2-methylpropionitrile, 3,5-Bis(2-cyano-2-methylethyl)toluene, 3,5-Bis(2-cyanoprop-2-yl)toluene
InChI Formula InChI=1S/C15H18N2/c1-11-6-12(14(2,3)9-16)8-13(7-11)15(4,5)10-17/h6-8H,1-5H3
SMILES Cc1cc(cc(c1)C(C)(C)C#N)C(C)(C)C#N
IUPAC Name 2-[3-(2-cyanopropan-2-yl)-5-methylphenyl]-2-methylpropanenitrile
Molecular Weight (g/mol) 226.32
Formula Weight 226.147
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.