missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ A 939572
GREENER_CHOICE

Product Code. 18716239
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18716239 10 mg 10mg
18721088 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18716239 Supplier Tocris Bioscience™ Supplier No. 4845/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent SCD-1 inhibitor

A 939572 is a potent stearoyl-CoA desaturase 1 (SCD1) inhibitor (IC50 = 0.4 nM). Selective for SCD1 over a range of kinases and hERG channels. Inhibits proliferation of squamous cell carcinoma FaDu cells in vitro. Also induces cell death of undifferentiated human embryonic stem cells (ESCs). Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(2-Chlorophenoxy)-N-[3-[(methylamino)carbonyl]phenyl]-1-piperidinecarboxamide
CAS 1032229-3-6
Quantity 10 mg
Target Compounds for Stem Cell Differentiation
Molecular Formula C20H22ClN3O3
Formula Weight 387.86
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.